missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenethyl bromide, 97%

Product Code. 10694165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694165 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10694165 Supplier Thermo Scientific Acros Supplier No. 398050050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 332-42-3 | C8H8BrF | 203.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 332-42-3
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Density 1.4498g/mL
Boiling Point 100°C to 104°C (15.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8BrF
Linear Formula FC6H4(CH2)2Br
Refractive Index 1.5310 to 1.5350 (20°C, 589nm)
MDL Number MFCD03844794
Quantity 5 g
Specific Gravity 1.4498
Synonym 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene
InChI Key FLRUNCJXOVYWDH-UHFFFAOYSA-N
SMILES FC1=CC=C(CCBr)C=C1
IUPAC Name 1-(2-bromoethyl)-4-fluorobenzene
Molecular Weight (g/mol) 203.05
PubChem CID 573153
Formula Weight 203.05
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Fluorophenethyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenethyl bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.