missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluorophenoxyacetic acid, 98+%

Product Code. 11304514
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304514 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11304514 Supplier Alfa Aesar™ Supplier No. L01216.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 405-79-8
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00004304
InChI Key ZBIULCVFFJJYTN-UHFFFAOYSA-N
Synonym 4-fluorophenoxyacetic acid, 2-4-fluorophenoxy acetic acid, p-fluorophenoxyacetic acid, 4-fluorophenoxy acetic acid, acetic acid, 4-fluorophenoxy, p-fluorophenoxy acetic acid, para-fluorophenoxyacetic acid, 4-fluorophenoxy acetic acid, alpha-carboxy-4-fluoroanisole, 2-p-fluorophenoxy acetic acid
PubChem CID 67882
IUPAC Name 2-(4-fluorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)F

Specifications

Melting Point 104°C to 105°C
Quantity 5 g
Beilstein 2259128
Formula Weight 170.14
Percent Purity ≥98%
Chemical Name or Material 4-Fluorophenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.