missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl isothiocyanate, 98%

Product Code. 11433017
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11433017

Brand: Thermo Scientific Alfa Aesar A15561.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1544-68-9 | C7H4FNS | 153.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1544-68-9
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00004809
InChI Key NFIUJHJMCQQYDL-UHFFFAOYSA-N
Synonym 4-fluorophenyl isothiocyanate, p-fluorophenyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanato, 4-fluorophenylisothiocyanate, 1-fluoro-4-isothiocyanato-benzene, isothiocyanic acid, p-fluorophenyl ester, isothiocyanic acid 4-fluorophenyl ester, 4-fluorobenzenisothiocyanate, 4-fluoroisothiocyanatobenzene, pubchem5008
PubChem CID 15241
IUPAC Name 1-fluoro-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)F

Specifications

Melting Point 24°C to 28°C
Density 1.25
Boiling Point 228°C
Flash Point 89°C (192°F)
Refractive Index 1.62
Quantity 5 g
UN Number UN2923
Beilstein 636596
Sensitivity Moisture sensitive
Formula Weight 153.18
Percent Purity 98%
Chemical Name or Material 4-Fluorophenyl isothiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenyl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluorophenyl isothiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.