missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetone, 99%

Product Code. 11352587
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352587

Brand: Thermo Scientific Alfa Aesar L11720.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 459-03-0 | C9H9FO | 152.17 g/mol

4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-03-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00000362
InChI Key ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Synonym 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one
PubChem CID 521187
IUPAC Name 1-(4-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(F)C=C1

Specifications

Density 1.11
Boiling Point 106°C to 107°C (18 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.4958
Quantity 5 g
Beilstein 1936697
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 152.17
Percent Purity 99%
Chemical Name or Material 4-Fluorophenylacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.