missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetyl chloride, 97%

Product Code. 11324106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324106 25 g 25g
11304106 1 g 1g
11314106 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324106 Supplier Thermo Scientific Alfa Aesar Supplier No. L19235.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 459-04-1 | C8H6ClFO | 172.58 g/mol

4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

Solubility
Miscible with dichloromethane.

Notes
Moisture Sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-04-1
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00674181
InChI Key SIOJFYRPBYGHOO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
PubChem CID 3744988
IUPAC Name 2-(4-fluorophenyl)acetyl chloride
SMILES FC1=CC=C(CC(Cl)=O)C=C1

Specifications

Density 1.259
Boiling Point 60°C (8 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.512
Quantity 25 g
UN Number UN3265
Beilstein 972148
Sensitivity Moisture sensitive
Solubility Information Miscible with dichloromethane.
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 4-Fluorophenylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.