missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetyl chloride, 98%

Product Code. 10177532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177532 1 g Glass bottle 1g
10013452 5 g Glass bottle 5g
10721145 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177532 Supplier Thermo Scientific Acros Supplier No. 376120010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 459-04-1 | C8H6ClFO | 172.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-04-1
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00674181
InChI Key SIOJFYRPBYGHOO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
PubChem CID 3744988
IUPAC Name 2-(4-fluorophenyl)acetyl chloride
SMILES FC1=CC=C(CC(Cl)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.2590g/mL
Boiling Point 60°C (8.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2C(O)Cl
Refractive Index 1.5110 to 1.513
Quantity 1 g
Specific Gravity 1.259
Solubility Information Solubility in water: reacts
Formula Weight 172.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorophenylacetyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenylacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.