missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetylene, 99%

Product Code. 15453437
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453437 5 g 5g
11386749 1 g 1g
11396749 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453437 Supplier Thermo Scientific Alfa Aesar Supplier No. H26269.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 766-98-3 | C8H5F | 120.126 g/mol

Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.

Solubility
Soluble in chloroform, diethyl ether, acetone, dichloromethane, hexane and ethanol. Highly soluble in toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-98-3
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.126
MDL Number MFCD00168823
InChI Key QXSWHQGIEKUBAS-UHFFFAOYSA-N
Synonym 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
PubChem CID 522627
IUPAC Name 1-ethynyl-4-fluorobenzene
SMILES C#CC1=CC=C(C=C1)F

Specifications

Melting Point 27°C to 28°C
Density 1.048
Boiling Point 55°C to 56°C (40 mmHg)
Flash Point 29°C (85°F)
Odor Characteristic
Refractive Index 1.517
Quantity 5 g
UN Number UN1325
Beilstein 1099285
Solubility Information Soluble in chloroform,diethyl ether,acetone,dichloromethane,hexane and ethanol. Highly soluble in toluene. Insoluble in water.
Formula Weight 120.12
Percent Purity 99%
Chemical Name or Material 4-Fluorophenylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.