missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylboronic acid, 97%, May contain varying amounts of anhydride, Maybridge

Product Code. 10359903
Change view
Click to view available options
:
Amber Glass Bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10359903 Amber Glass Bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10359903 Supplier Thermo Scientific Maybridge Supplier No. AC33410DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1765-93-1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H6BFO2
MDL Number MFCD00039136
Quantity 5 g
Synonym 4-fluorobenzeneboronic acid, 4-fluorophenyl boronic acid, 4-fluorophenyl boranediol, p-fluorophenylboronic acid, 4-fluorophenylbornic acid, 4-fluorophenyl dihydroxyboron, 4-fluorophenyl dihydroxyborane, p-fluorophenyl boric acid, 4-fluorophenyl boric acid
InChI Key LBUNNMJLXWQQBY-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(F)C=C1
IUPAC Name (4-fluorophenyl)boronic acid
Molecular Weight (g/mol) 139.92
PubChem CID 285645
ChEBI CHEBI:48661
Formula Weight 139.92
Chemical Name or Material 4-Fluorophenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.