missing translation for 'onlineSavingsMsg'
Learn More

(4-Fluorophenylthio)acetic acid, 97%

Product Code. 11404207
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404207 1 g 1g
11414207 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404207 Supplier Thermo Scientific Alfa Aesar Supplier No. A19658.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 332-51-4 | C8H7FO2S | 186.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-51-4
Molecular Formula C8H7FO2S
Molecular Weight (g/mol) 186.2
MDL Number MFCD00219825
InChI Key HQEROVXUKINLPI-UHFFFAOYSA-N
Synonym 4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
PubChem CID 736197
IUPAC Name 2-(4-fluorophenyl)sulfanylacetic acid
SMILES C1=CC(=CC=C1F)SCC(=O)O

Specifications

Melting Point 76°C to 79°C
Quantity 1 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material (4-Fluorophenylthio)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.