missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylurea, 96%

Product Code. 11344554
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344554 5 g 5g
11354554 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344554 Supplier Thermo Scientific Alfa Aesar Supplier No. L01314.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 659-30-3 | C7H7FN2O | 154.14 g/mol

4-Fluorophenylurea is widely used as a filter for drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylurea is widely used as a filter for drug.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 659-30-3
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00014787
InChI Key IQZBVVPYTDHTIP-UHFFFAOYSA-N
Synonym 4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea
PubChem CID 12612
IUPAC Name (4-fluorophenyl)urea
SMILES NC(=O)NC1=CC=C(F)C=C1

Specifications

Melting Point 183°C to 185°C
Quantity 5 g
Beilstein 2090131
Solubility Information Slightly soluble in water.
Formula Weight 154.14
Percent Purity 96%
Chemical Name or Material 4-Fluorophenylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.