missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalic anhydride, 98%, Thermo Scientific™

Product Code. 11439287
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11439287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21352.03
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

Solubility
Soluble in sodium hydroxide.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 319-03-9
Melting Point 74°C to 79°C
Density 1.5
Boiling Point 147°C to 149°C (20 mmHg)
Molecular Formula C8H3FO3
MDL Number MFCD00191363
Quantity 1 g
Beilstein 131281
Sensitivity Moisture sensitive
Synonym 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride
Solubility Information Soluble in sodium hydroxide.
InChI Key XVMKZAAFVWXIII-UHFFFAOYSA-N
SMILES FC1=CC=C2C(=O)OC(=O)C2=C1
IUPAC Name 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 166.11
PubChem CID 67572
Formula Weight 166.11
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalic anhydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.