missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorothiophenol, 97%

Product Code. 11448227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448227 100 g 100g
11428227 5 g 5g
11438227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448227 Supplier Thermo Scientific Alfa Aesar Supplier No. A17600.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 371-42-6 | C6H5FS | 128.164 g/mol

It is used in the synthesis of Bicalutamide, an anti-cancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of Bicalutamide, an anti-cancer drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-42-6
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.164
MDL Number MFCD00004846
InChI Key OKIHXNKYYGUVTE-UHFFFAOYSA-N
Synonym 4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
PubChem CID 67789
IUPAC Name 4-fluorobenzenethiol
SMILES C1=CC(=CC=C1F)S

Specifications

Density 1.203
Boiling Point 170°C to 172°C
Flash Point 57°C (135°F)
Odor Unpleasant
Refractive Index 1.55
Quantity 100 g
UN Number UN3071
Beilstein 1446350
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.17
Percent Purity ≥98%
Assay (dry wt.), water <7%
Chemical Name or Material 4-Fluorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.