missing translation for 'onlineSavingsMsg'
Learn More

4-(Fmoc-amino)benzoic acid, 97%

Product Code. 15499597
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499597 250 mg 250mg
15479597 1 g 1g
15489597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52828.MD
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 185116-43-2 | C22H17NO4 | 359.381 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185116-43-2
Molecular Formula C22H17NO4
Molecular Weight (g/mol) 359.381
MDL Number MFCD00144888
InChI Key VGSYYBSAOANSLR-UHFFFAOYSA-N
Synonym fmoc-4-abz-oh, fmoc-paba-oh, n-fmoc-4-aminobenzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, ambotzfaa1645, fmoc-p-aminobenzoic acid, 4-fmoc-amino benzoic acid, chembl46378
PubChem CID 2756082
IUPAC Name 4-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=C(C=C4)C(=O)O

Specifications

Quantity 250 mg
Beilstein 7722986
Formula Weight 359.38
Percent Purity 97%
Chemical Name or Material 4-(Fmoc-amino)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.