missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzene-1-sulfonamide hydrochloride, Maybridge™

Product Code. 10354412
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10354412 10 g 10g
10262912 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354412 Supplier Thermo Scientific Maybridge Supplier No. BTB06645EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17852-52-7
Molecular Formula C6H10ClN3O2S
Molecular Weight (g/mol) 223.675
MDL Number MFCD00052262
InChI Key IKEURONJLPUALY-UHFFFAOYSA-N
Synonym 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride
PubChem CID 2794567
IUPAC Name 4-hydrazinylbenzenesulfonamide;hydrochloride
SMILES C1=CC(=CC=C1NN)S(=O)(=O)N.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula NH2NHC6H4SO2NH2·HCl
Quantity 10 g
Formula Weight 223.68
Percent Purity 95%
Chemical Name or Material 4-Hydrazinobenzene-1-sulfonamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.