missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonic acid hydrate, 98%

Product Code. 11460993
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460993 100 g 100g
11440993 5 g 5g
11450993 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460993 Supplier Thermo Scientific Alfa Aesar Supplier No. A13362.22
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-71-5 | C6H8N2O3S | 188.20 g/mol

4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

Solubility
It is soluble in water.

Notes
Stable under normal conditions.Keep away from strong oxidizing agents. Store in closed vessels.Hygroscopic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-71-5
Molecular Formula C6H8N2O3S
Molecular Weight (g/mol) 188.20
MDL Number MFCD00025097
InChI Key IOMZCWUHFGMSEJ-UHFFFAOYSA-N
Synonym 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino
PubChem CID 66825
IUPAC Name 4-hydrazinylbenzenesulfonic acid
SMILES NNC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Melting Point ∼286°C (decomposition)
Quantity 100 g
Beilstein 514779
Sensitivity Hygroscopic
Merck Index 14,4773
Solubility Information It is soluble in water.
Formula Weight 188.20 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Hydrazinobenzenesulfonic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.