missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid, 97%

Product Code. 11441558
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441558 250 g 250g
11421558 10 g 10g
11431558 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441558 Supplier Thermo Scientific Alfa Aesar Supplier No. A13874.30
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-67-0 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-67-0
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007581
InChI Key PCNFLKVWBDNNOW-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911
PubChem CID 12089
IUPAC Name 4-hydrazinylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)NN

Specifications

Melting Point ∼216°C (decomposition)
Quantity 250 g
Beilstein 387378
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 4-Hydrazinobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.