missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid hydrochloride, 97%

Product Code. 15119323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15119323 10 g 10g
15139323 50 g 50g
15149323 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15119323 Supplier Thermo Scientific Acros Supplier No. 450670100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24589-77-3 | C7H9ClN2O2 | 188.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-77-3
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.61
MDL Number MFCD00039073
InChI Key XHLQMKQBCHYRLC-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride
PubChem CID 90547
IUPAC Name 4-hydrazinylbenzoic acid;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 252.0°C to 254.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 188.61
Percent Purity 97%
Chemical Name or Material 4-Hydrazinobenzoic acid hydrochloride
compliance-icons
Product Identifier
  • 4-Hydrazinobenzoic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.