missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid hydrochloride, 98%

Product Code. 11489846
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489846 10 g 10g
11499846 50 g 50g
11409856 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489846 Supplier Thermo Scientific Alfa Aesar Supplier No. B20326.09
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24589-77-3 | C7H9ClN2O2 | 188.61 g/mol

4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-77-3
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.61
MDL Number MFCD00039073
InChI Key XHLQMKQBCHYRLC-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride
PubChem CID 90547
IUPAC Name 4-hydrazinylbenzoic acid;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 252°C to 254°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 188.61
Percent Purity 98%
Chemical Name or Material 4-Hydrazinobenzoic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.