missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,3,5,6-tetrafluorobenzoic acid, 97%

Product Code. p-7048083
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11329056

Brand: Thermo Scientific Alfa Aesar L20209.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 652-34-6 | C7H2F4O3 | 210.084 g/mol

2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid hydrate may be used in the preparation of new ligand, methyl 2,3,5,6-tetrafluoro-4-oxybenzoate (C8H3F4O3), combining an electron withdrawing group (C6F4) to tune the reactivity with an anchor group (CO2Me) for immobilization on supports.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid hydrate may be used in the preparation of new ligand, methyl 2,3,5,6-tetrafluoro-4-oxybenzoate (C8H3F4O3), combining an electron withdrawing group (C6F4) to tune the reactivity with an anchor group (CO2Me) for immobilization on supports.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-34-6
Molecular Formula C7H2F4O3
Molecular Weight (g/mol) 210.084
MDL Number MFCD00129952
InChI Key FTLHGQOBAPTEHE-UHFFFAOYSA-N
Synonym 4-hydroxy-2,3,5,6-tetrafluorobenzoic acid, 2,3,5,6-tetrafluoro-4-hydroxy-benzoic acid, tetrafluoro-4-hydroxybenzoic acid, benzoic acid, 2,3,5,6-tetrafluoro-4-hydroxy, f4-pohb, tetrafluoro-4-hydroxybenzoate, zlchem 412, acmc-1b1y5, ksc622i2b, 4-carboxy-2,3,5,6-tetrafluorophenol
PubChem CID 98835
IUPAC Name 2,3,5,6-tetrafluoro-4-hydroxybenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)O)F)F)C(=O)O

Specifications

Melting Point 148°C to 150°C (decomposition)
Quantity 1 g
Beilstein 986011
Solubility Information Insoluble in water.
Formula Weight 210.1
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-2,3,5,6-tetrafluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.