missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-2-methylbenzeneboronic acid, 98%

Product Code. 15469837
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469837 250 mg 250mg
15459837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469837 Supplier Alfa Aesar™ Supplier No. H53068.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 493035-82-8
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD03788424
InChI Key OYIYNIONWDBJIF-UHFFFAOYSA-N
Synonym 4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
PubChem CID 2773448
IUPAC Name (4-hydroxy-2-methylphenyl)boronic acid
SMILES CC1=CC(O)=CC=C1B(O)O

Specifications

Melting Point 100°C to 104°C
Quantity 250 mg
Formula Weight 151.96
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2-methylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.