missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%

Product Code. 15209946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15209946 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15209946 Supplier Thermo Scientific Acros Supplier No. 454680100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 81-81-2 | C19H16O4 | 308.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81-81-2
Melting Point 162.0°C to 164.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C19H16O4
Quantity 10 g
Merck Index 15,10236
Synonym warfarin, coumadin, coumafene, prothromadin, coumafen, coumarins, zoocoumarin, coumefene, panwarfin, warfarine
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, dioxane, moderatly soluble in, methanol, ethanol, isopropanol, insoluble in benzene, cyclohexane
InChI Key PJVWKTKQMONHTI-UHFFFAOYSA-N
SMILES CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
Molecular Weight (g/mol) 308.34
PubChem CID 54678486
ChEBI CHEBI:87732
Formula Weight 308.34
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • H360D-May damage the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.