missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzonitrile, 97%

Product Code. 11411658
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411658 25 g 25g
11401658 5 g 5g
11421658 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411658 Supplier Thermo Scientific Alfa Aesar Supplier No. A13929.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1689-83-4 | C7H3I2NO | 370.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1689-83-4
Molecular Formula C7H3I2NO
Molecular Weight (g/mol) 370.92
MDL Number MFCD00016422
InChI Key NRXQIUSYPAHGNM-UHFFFAOYSA-N
Synonym ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox
PubChem CID 15530
ChEBI CHEBI:81821
IUPAC Name 4-hydroxy-3,5-diiodobenzonitrile
SMILES OC1=C(I)C=C(C=C1I)C#N

Specifications

Melting Point 200°C to 207°C
Odor Odorless
Quantity 25 g
UN Number UN3439
Beilstein 2364041
Sensitivity Light sensitive
Formula Weight 370.92
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-diiodobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.