missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxycinnamic acid, 98%

Product Code. 11467617
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467617 25 g 25g
11379519 1 g 1g
11457617 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467617 Supplier Thermo Scientific Alfa Aesar Supplier No. A15676.14
 Restricted Item
Estimated Shipment: 25-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 530-59-6 | C11H12O5 | 224.212 g/mol

4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

Solubility
Soluble in methanol, ethanol and dimethyl sulfoxide.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD00004401
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Spezifikation

Melting Point ∼195°C (decomposition)
Quantity 25 g
Beilstein 2130006
Solubility Information Soluble in methanol,ethanol and dimethyl sulfoxide.
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxycinnamic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.