missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzoic acid, 98%

Product Code. 11449993
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449993 1 g 1g
11459993 5 g 5g
11469993 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449993 Supplier Thermo Scientific Alfa Aesar Supplier No. A13051.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4919-37-3 | C9H10O3 | 166.176 g/mol

4-hydroxy-3,5-dimethylbenzoic acid is widely used as a reagent for potent TTR amyloidogenesis inhibitors. It plays an important role as an intermediate in agrochemicals and dyestuffs. It is also used in the preparation of mexiletine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-hydroxy-3,5-dimethylbenzoic acid is widely used as a reagent for potent TTR amyloidogenesis inhibitors. It plays an important role as an intermediate in agrochemicals and dyestuffs. It is also used in the preparation of mexiletine derivatives.

Solubility
Soluble in methanol.

Notes
Keep away from heat and avoid excessive light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4919-37-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016536
InChI Key OMNHTTWQSSUZHO-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzoic acid, benzoic acid, 4-hydroxy-3,5-dimethyl, pubchem16247, acmc-209kf7, ksc496e2b, omnhttwqssuzho-uhfffaoysa, 3.5-dimethyl-4-hydroxybenzoic acid, 3,5-dimethyl-4-hydroxy-benzoic acid, 4-hydroxy-3,5-dimethyl-benzoic acid
PubChem CID 138387
IUPAC Name 4-hydroxy-3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1O)C)C(=O)O

Specifications

Melting Point 221°C to 226°C
Quantity 1 g
Beilstein 2576289
Solubility Information Soluble in methanol.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzoic acid
compliance-icons
Product Identifier
  • 4-Hydroxy-3,5-dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hydroxy-3,5-dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.