missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dinitrobenzoic acid, 98+%

Product Code. 11410657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410657 1 g 1g
11420657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22474.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1019-52-9 | C7H4N2O7 | 228.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019-52-9
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.12
MDL Number MFCD00017000
InChI Key GBSWIDSKAJFWMF-UHFFFAOYSA-N
Synonym 3,5-dinitro-4-hydroxybenzoic acid, acmc-20anc3, 4-hydroxy-3,5-dinitro-benzoic acid, #, benzoic acid,4-hydroxy-3,5-dinitro, 4-hydroxy-3,5-dinitrobenzoic acid, 98+%
PubChem CID 70552
IUPAC Name 4-hydroxy-3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 248°C to 252°C
Quantity 1 g
Formula Weight 228.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.