missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dinitrobenzoic acid, 98+%

Product Code. p-7026468
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11410657

Brand: Thermo Scientific Alfa Aesar B22474.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1019-52-9 | C7H4N2O7 | 228.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019-52-9
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.12
MDL Number MFCD00017000
InChI Key GBSWIDSKAJFWMF-UHFFFAOYSA-N
Synonym 3,5-dinitro-4-hydroxybenzoic acid, acmc-20anc3, 4-hydroxy-3,5-dinitro-benzoic acid, #, benzoic acid,4-hydroxy-3,5-dinitro, 4-hydroxy-3,5-dinitrobenzoic acid, 98+%
PubChem CID 70552
IUPAC Name 4-hydroxy-3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 248°C to 252°C
Quantity 1 g
Formula Weight 228.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.