missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dinitrophenylacetic acid, 97%, Thermo Scientific™

Product Code. 11322547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322547 5 g 5g
11312547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322547 Supplier Thermo Scientific Alfa Aesar Supplier No. L11643.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10463-37-3
Molecular Formula C8H6N2O7
Molecular Weight (g/mol) 242.143
MDL Number MFCD00016995
InChI Key MLVYQQLUGFSXQH-UHFFFAOYSA-N
Synonym 3,5-dinitro-4-hydroxyphenylacetic acid, 4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrophenylacetic acid, 3,5-dinitro-4-hydroxyphenacetic acid, 4-hydroxy-3,5-dinitro-phenyl-acetic acid, 2-4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrobenzeneacetic acid, 4-hydroxy-3,5-dinitrophenacetyl, acmc-20anc2, 3,5-dinitro-4-hydroxyphenyl-acetic acid
PubChem CID 98596
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 166°C to 176°C
Quantity 5 g
Beilstein 2665595
Formula Weight 242.15
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-dinitrophenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.