missing translation for 'onlineSavingsMsg'
Learn More

4-hydroxy-3-iodo-5-methoxybenzaldehyde, Thermo Scientific™

Product Code. p-4391363
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10535143

Brand: Thermo Scientific Maybridge SB01117EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5438-36-8
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
InChI Key FBBCSYADXYILEH-UHFFFAOYSA-N
Synonym 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
PubChem CID 79499
IUPAC Name 4-hydroxy-3-iodo-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)I)O

Specifications

Assay Percent Range 97%
Linear Formula IC6H2(OCH3)(OH)CHO
Quantity 10 g
Formula Weight 278.04
Percent Purity 97%
Chemical Name or Material 5-Iodovanillin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.