missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-3-methoxybenzyl alcohol, 98+%

Product Code. 11462148
Change view
Click to view available options
Quantity:
50g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462148 50g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462148 Supplier Alfa Aesar™ Supplier No. A15160.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Used as a flavoring agent. Other possible uses, vanilla, coconut, cream and other dairy nuances, coumarin.

Solubility
Soluble in 95% ethanol (5 %), water, and oils.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 498-00-0
Molecular Formula C8H10O3
MDL Number MFCD00004659
Quantity 50g
Beilstein 1910044
Synonym vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol
InChI Key ZENOXNGFMSCLLL-UHFFFAOYSA-N
SMILES COC1=CC(CO)=CC=C1O
IUPAC Name 4-(hydroxymethyl)-2-methoxyphenol
Molecular Weight (g/mol) 154.17
PubChem CID 62348
ChEBI CHEBI:18353
Formula Weight 154.17
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.