missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzyl alcohol, 99%

Product Code. 10337690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10337690 50 g Plastic bottle 50g
10504121 250 g Plastic bottle 250g
13408428 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10337690 Supplier Thermo Scientific Acros Supplier No. 174580500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 498-00-0 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-00-0
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00004659
InChI Key ZENOXNGFMSCLLL-UHFFFAOYSA-N
Synonym vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol
PubChem CID 62348
ChEBI CHEBI:18353
IUPAC Name 4-(hydroxymethyl)-2-methoxyphenol
SMILES COC1=CC(CO)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 112.0°C to 115.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H3(OCH3)CH2OH
Quantity 50 g
Beilstein 06,IV,7381
Formula Weight 154.16
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.