missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%

Product Code. 11439077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439077 25 g 25g
11419077 1 g 1g
11429077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439077 Supplier Thermo Scientific Alfa Aesar Supplier No. B20637.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7149-10-2 | C8H11ClNO2 | 188.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-2-7149
MDL Number MFCD00012864

Specifications

Melting Point ∼220°C (decomposition)
Molecular Formula C8H11ClNO2
Quantity 25 g
Beilstein 3915418
Sensitivity Moisture sensitive
Synonym 4-hydroxy-3-methoxybenzylamine hydrochloride, 4-aminomethyl-2-methoxyphenol hydrochloride, vanillylamine hydrochloride, n-vanillylamine hydrochloride, 2-vanillylamine hydrochloride, 4-hydroxy-3-methoxybenzylamine hcl, phenol, 4-aminomethyl-2-methoxy-, hydrochloride, 4-aminomethyl-2-methoxyphenol, chloride, vanillylamine hcl
InChI Key LOYPVODLNGWOLM-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)CN)O.Cl
IUPAC Name 4-(aminomethyl)-2-methoxyphenol;hydrochloride
Molecular Weight (g/mol) 188.63
PubChem CID 165576
Formula Weight 189.64
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.