missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzaldehyde, 97%

Product Code. p-146603 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10104180

Brand: Thermo Scientific Acros 162890050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3011-34-5 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3011-34-5
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007117
InChI Key YTHJCZRFJGXPTL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
PubChem CID 18169
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 144°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(NO2)CHO
Quantity 5 g
Beilstein 08, 83
Formula Weight 167.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-3-nitrobenzaldehyde, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.