missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzonitrile 97%, ACROS Organics™

Product Code. 10404702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10404702 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10404702 Supplier Thermo Scientific Acros Supplier No. 344040100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3272-08-0
Molecular Formula C7H3N2O3
Linear Formula NO2C6H3(OH)CN
MDL Number MFCD00070775
Quantity 10 g
Synonym 4-hydroxy-3-nitrobenzonitrile, 2-nitro-4-cyanophenol, benzonitrile, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzonitrile, 4-cyano-2-nitrophenol, 4-hydroxy-3-nitro-benzonitrile, 4-hydroxy-3-nitrobenzenecarbonitrile, pubchem19690, acmc-1cq5c, ksc494q7b
InChI Key INBLGVOPOSGVTA-UHFFFAOYSA-M
SMILES [O-]C1=CC=C(C=C1[N+]([O-])=O)C#N
IUPAC Name 4-cyano-2-nitrobenzen-1-olate
Molecular Weight (g/mol) 163.11
PubChem CID 76758
Formula Weight 164.12
Chemical Name or Material 4-Hydroxy-3-nitrobenzonitrile, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.