missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrocoumarin, 98%

Product Code. 11424903
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424903 25 g 25g
11414903 5 g 5g
11434903 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424903 Supplier Thermo Scientific Alfa Aesar Supplier No. A12221.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20261-31-8 | C9H5NO5 | 207.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 20261-31-8
Molecular Formula C9H5NO5
Molecular Weight (g/mol) 207.141
MDL Number MFCD00051671
InChI Key NZQAQAUWFHMVEM-UHFFFAOYSA-N
PubChem CID 54682640
IUPAC Name 4-hydroxy-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O

Spezifikation

Melting Point ∼171°C (decomposition)
Quantity 25 g
Beilstein 1247343
Synonym 4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
Formula Weight 207.14
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-nitrocoumarin

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.