missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%

Product Code. 11491518
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491518 25 g 25g
11471518 1 g 1g
11481518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491518 Supplier Thermo Scientific Alfa Aesar Supplier No. A13858.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10463-20-4 | C8H7NO5 | 197.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10463-20-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007122
InChI Key QBHBHOSRLDPIHG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
PubChem CID 447364
ChEBI CHEBI:546274
SMILES C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O

Specifications

Melting Point 146°C to 148°C
Quantity 25 g
Beilstein 2696044
Formula Weight 197.15
Percent Purity 99%
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.