missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methylcoumarin, 98+%

Product Code. 10536793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10536793 5 g Glass bottle 5g
10169122 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10536793 Supplier Thermo Scientific Acros Supplier No. 372870050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13252-83-0 | C10H8O3 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13252-83-0
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00085738
InChI Key CSVFZDYJGBVNEX-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methylcoumarin, 4-hydroxy-6-methyl-2h-chromen-2-one, 6-methyl-4-hydroxycoumarin, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2-hydroxy-6-methylchromen-4-one, 2-hydroxy-6-methyl-4h-chromen-4-one, 6-methyl-4-hydroxy coumarin, hydroxy-6-methylcoumarin,4
PubChem CID 54686349
IUPAC Name 4-hydroxy-6-methylchromen-2-one
SMILES CC1=CC=C2OC(O)=CC(=O)C2=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 261.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in hot ethanol and methanol
Formula Weight 176.17
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-6-methylcoumarin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.