missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Kresoxim-methyl Carboxylic Acid, TRC

Product Code. 30406112
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406112 10 mg
30402781 100 mg
30396945 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406112 Supplier Toronto Research Chemicals Supplier No. TRCH94369010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181373-11-5
Molecular Formula C17 H17 N O5
Molecular Weight (g/mol) 315.32
Synonym 2-[(4-Hydroxy-2-methylphenoxy)methyl]-α-(methoxyimino)benzeneacetic acid
IUPAC Name 2-[2-[(4-hydroxy-2-methylphenoxy)methyl]phenyl]-2-methoxyiminoacetic acid
SMILES CON=C(C(=O)O)c1ccccc1COc2ccc(O)cc2C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17NO5/c1-11-9-13(19)7-8-15(11)23-10-12-5-3-4-6-14(12)16(17(20)21)18-22-2/h3-9,19H,10H2,1-2H3,(H,20,21)
Formula Weight 315.1107
Chemical Name or Material (EZ)-Kresoxim-4-hydroxy (free acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.