missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone, 98%

Product Code. 11497397
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497397 1000 g 1000g
11407407 50 g 50g
11417407 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497397 Supplier Thermo Scientific Alfa Aesar Supplier No. A15096.0B
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1137-42-4 | C13H10O2 | 198.22 g/mol

4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002355
InChI Key NPFYZDNDJHZQKY-UHFFFAOYSA-N
Synonym 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
PubChem CID 14347
ChEBI CHEBI:34421
IUPAC Name (4-hydroxyphenyl)-phenylmethanone
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 132°C to 135°C
Boiling Point 260°C to 262°C (24 mmHg)
Quantity 1000 g
Beilstein 777776
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.