missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxybiphenyl-4-carboxylic acid, 99%

Product Code. 11455047
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455047 2.5 g 2.5g
11465047 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455047 Supplier Thermo Scientific Alfa Aesar Supplier No. B21946.88
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58574-03-1 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58574-03-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00059078
InChI Key JTGCXYYDAVPSFD-UHFFFAOYSA-N
Synonym 4'-hydroxy-4-biphenylcarboxylic acid, 4-4-hydroxyphenyl benzoic acid, 4'-hydroxybiphenyl-4-carboxylic acid, 4'-hydroxy-1,1'-biphenyl-4-carboxylic acid, 4-hydroxy-4'-biphenylcarboxylic acid, 4'-hydroxy biphenyl-4-carboxylic acid, 4-hydroxybiphenyl-4'-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-hydroxy, chembl90522, 4'-hydroxy 1,1'-biphenyl-4-carboxylic acid
PubChem CID 301556
IUPAC Name 4-(4-hydroxyphenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)C(=O)O

Specifications

Melting Point 292°C to 295°C
Quantity 2.5 g
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material 4'-Hydroxybiphenyl-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.