missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyhippuric Acid, TRC

Product Code. 30009554
Change view
Click to view available options
Quantity:
25 mg
50 mg
1 g
Unit Size:
1g
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009554 25 mg 25mg
30005706 1 g 1g
30003636 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009554 Supplier Toronto Research Chemicals Supplier No. TRCH82781025MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2482-25-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
Synonym N-(4-Hydroxybenzoyl)glycine, 4-Hydroxybenzoylglycine, N-(p-Hydroxybenzoyl)glycine, p-Hydroxyhippuric Acid
IUPAC Name 2-[(4-hydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)c1ccc(O)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H9NO4/c11-7-3-1-6(2-4-7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)
Formula Weight 195.0532
Chemical Name or Material 4-Hydroxyhippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.