missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyindole, 98%

Product Code. 15479009
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479009 1 g 1g
15489009 5 g 5g
15499009 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15479009 Supplier Thermo Scientific Alfa Aesar Supplier No. L14352.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2380-94-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-94-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005667
InChI Key NLMQHXUGJIAKTH-UHFFFAOYSA-N
Synonym 4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
PubChem CID 75421
ChEBI CHEBI:24702
IUPAC Name 1H-indol-4-ol
SMILES OC1=C2C=CNC2=CC=C1

Specifications

Melting Point 97°C to 99°C
Quantity 1 g
Beilstein 114905
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxyindole
compliance-icons
Product Identifier
  • 4-Hydroxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hydroxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.