accessibility menu, dialog, popup

UserName

CAS RN 2380-94-1

OHHN

CAS RN 2380-94-1

IUPAC Name: 1H-indol-4-ol
Synonyms: hydroxyindole
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: NLMQHXUGJIAKTH-UHFFFAOYSA-N
SMILES: OC1=C2C=CNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results