missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenoxyacetic acid, 98+%, Thermo Scientific™

Product Code. 11332757
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332757 250 mg 250mg
11322757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332757 Supplier Thermo Scientific Alfa Aesar Supplier No. L15721.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68858-21-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00057827
InChI Key VUCNQOPCYRJCGQ-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenoxyacetic acid, 4-hydroxymethylphenoxyacetic acid, 2-4-hydroxymethyl phenoxy acetic acid, hmp linker, p-hydroxymethyl phenoxyacetic acid, 4-hydroxymethyl-phenoxy acetic acid
PubChem CID 194291
IUPAC Name 2-[4-(hydroxymethyl)phenoxy]acetic acid
SMILES C1=CC(=CC=C1CO)OCC(=O)O

Specifications

Melting Point 110°C to 113°C
Quantity 250 mg
Beilstein 5260336
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 4-(Hydroxymethyl)phenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.