missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The 4-Hydroxynonenal antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na 4-Hydroxynonenal. This antibody recognizes 4-hydroxynonenal antigen. The 4-Hydroxynonenal antibody has been validated for the following applications: ELISA analysis.
Specifications
Specifications
| Antigen | 4-Hydroxynonenal |
| Applications | ELISA |
| Clone | 2C1D10 |
| Concentration | 1000 μg/mL |
| Gene Symbols | 4-HNE, 4-Hydroxy-2-nonenal, 4-Hydroxynonenal, C9H16O2, Cas. 75899-68-2 |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Antibody Molecular Weight | 150 kDa |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?