missing translation for 'onlineSavingsMsg'
Learn More

(4-Hydroxyphenoxy)acetic acid, 98%, Thermo Scientific Chemicals

Product Code. 10280023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10280023 5 g Glass bottle 5g
10675681 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10280023 Supplier Thermo Scientific Acros Supplier No. 412090050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-84-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00014362
InChI Key PKGWLCZTTHWKIZ-UHFFFAOYSA-N
Synonym 4-hydroxyphenoxyacetic acid, 4-hydroxyphenoxy acetic acid, 2-4-hydroxyphenoxy acetic acid, 4-hydroxypheoxyacetate, acetic acid, 4-hydroxyphenoxy, 4-hydroxy-phenoxy-acetic acid, p-hydroxyphenoxyacetic acid, acetic acid, 2-4-hydroxyphenoxy, acetic acid, p-hydroxyphenoxy, 4-carboxymethoxy phenol
PubChem CID 15881
ChEBI CHEBI:1881
IUPAC Name 2-(4-hydroxyphenoxy)acetic acid
SMILES C1=CC(=CC=C1O)OCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 152°C to 156°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 168.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material (4-Hydroxyphenoxy)acetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (4-Hydroxyphenoxy)acetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.