missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl benzoate, 98%

Product Code. 15565166 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15565166 25 g 25g
15555166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15565166 Supplier Thermo Scientific Acros Supplier No. 455730250
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2444-19-1 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-19-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00053304
InChI Key JFAXJRJMFOACBO-UHFFFAOYSA-N
Synonym hydroquinone monobenzoate, p-hydroxyphenyl benzoate, unii-zcy7o225cp, 4-benzoyloxyphenol, 1,4-diphenol, 1-benzoate, benzoic acid 4-hydroxyphenyl ester, 4-hydroxyphenyl benzoate, zcy7o225cp, 4-hydroxyphenylbenzoate, 4-hydroxyphenylbenzoat
PubChem CID 75549
IUPAC Name (4-hydroxyphenyl) benzoate
SMILES OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 164.0°C to 165.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 214.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphenyl benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.