missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetamide, 99%, Thermo Scientific™

Product Code. 10570522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10570522 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10570522 Supplier Thermo Scientific Acros Supplier No. 254270050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 17194-82-0
CAS Min % 98.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9NO2
MDL Number MFCD00017145
Quantity 5 g
Beilstein 10, III, 434
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,methanol
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CC(=O)N)O
IUPAC Name 2-(4-hydroxyphenyl)acetamide
Molecular Weight (g/mol) 151.16
PubChem CID 86986
Formula Weight 151.16
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxyphenylacetamide, 99%
Show More Show Less
compliance-icons
Produktidentifikator
  • 4-Hydroxyphenylacetamide
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.