missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetamide, 99%

Product Code. 11482318
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482318 10 g 10g
11492318 50 g 50g
11402328 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482318 Supplier Thermo Scientific Alfa Aesar Supplier No. A15266.09
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17194-82-0 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00017145
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O

Specifications

Melting Point 173°C to 175°C
Quantity 10 g
Beilstein 2085984
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 4-Hydroxyphenylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.