missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylpyruvic Acid, TRC

Product Code. 17954185
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17954185 Supplier Toronto Research Chemicals Supplier No. TRCH949870100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 156-39-8
Molecular Formula C9H8O4
Quantity 100 mg
Synonym (p-Hydroxyphenyl)pyruvic Acid, 3-(4-Hydroxyphenyl)-2-oxopropanoic Acid; 3-(4-Hydroxyphenyl)-2-oxopropionic Acid, 3-(4-Hydroxyphenyl)pyruvic Acid; 3-(p-Hydroxyphenyl)-2-oxopropionic Acid, 3-(p-Hydroxyphenyl)pyruvic Acid; 4-Hydroxy-α-oxobenzenepropanoic Acid, HPPA, NSC 100738, NSC 666757; Testacid
InChI Formula InChI=1S/C9H8O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,10H,5H2,(H,12,13)
SMILES OC(=O)C(=O)Cc1ccc(O)cc1
IUPAC Name 3-(4-hydroxyphenyl)-2-oxopropanoic acid
Molecular Weight (g/mol) 180.16
Formula Weight 180.0423
Chemical Name or Material 4-Hydroxyphenylpyruvic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.