missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphthalic acid, 98%

Product Code. 11487397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487397 100 g 100g
11467397 5 g 5g
11477397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487397 Supplier Thermo Scientific Alfa Aesar Supplier No. A15091.22
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 610-35-5 | C8H6O5 | 182.131 g/mol

4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-35-5
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00013984
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
PubChem CID 11881
ChEBI CHEBI:27600
IUPAC Name 4-hydroxyphthalic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 100 g
Solubility Information Slightly miscible with water.
Formula Weight 182.13
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.